Tri-​O-​benzoyl-​D-​galactal 98%

Supplier: AFG Bioscience

Synonyms: (2R,3R,4R)-2-((Benzoyloxy)methyl)-3,4-dihydro-2H-pyran-3,4-diyl dibenzoate

C53316-10G C53316-5G
76619-892EA 545.18 CAD
76619-892 76619-890
Tri-​O-​benzoyl-​D-​galactal 98%
Tri-​O-​benzoyl-​D-​galactal
Formula: C₂₇H₂₂O₇
MW: 458.45 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00135160
CAS Number: 34948-79-3

Order Now

Specification Test Results

Purity Limit 98%
Appearance White powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR