Phenyl 1-thio-β-D-galactopyranoside ≥95%

Supplier: AFG Bioscience

C53296-1G C53296-5G
76619-880EA 155.76 CAD
76619-880 76619-882
Phenyl 1-thio-β-D-galactopyranoside ≥95%
Phenyl 1-thio-β-D-galactopyranoside
Formula: C₁₂H₁₆O₅S
Storage Temperature: Refrigerator
MDL Number: MFCD00063272
CAS Number: 16758-34-2

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White To Pale Yellow Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR