Methyl N-acetyl-α-D-glucosaminide 95%
Supplier: AFG Bioscience
Synonyms:
Methyl 2-acetamido-2-deoxy-alpha-d-glucopyranoside, N-((2S,3R,4R,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-2-methoxytetrahydro-2H-pyran-3-yl)acetamide, Methyl 2-acetamido-2-deoxy-α-D-glucopyranoside
C54049-500MG
C54049-1G
C54049-2G
C54049-250MG
76620-010EA
155.76
CAD
76620-010
76620-012
76620-014
77888-971
Methyl N-acetyl-α-D-glucosaminide 95%
Methyl N-acetyl-α-D-glucosaminide
Formula:
C₉H₁₇NO₆ MW: 235.23 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00056076 CAS Number: 6082-04-8 |
Specification Test Results
Purity Limit | 95% |
Appearance | white powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...