UDP-Xylose disodium salt 97%

Supplier: AFG Bioscience

Synonyms: UDP-xylose disodium, Uridine 5'-(trihydrogen diphosphate),P'-α-D-xylopyranosyl ester, disodium salt, Uridine[5']diphospho-a-D-xylopyranoside disodium

C55201-10MG C55201-5MG
76620-176EA 77.88 CAD
76620-176 76620-174
UDP-Xylose disodium salt 97%
UDP-Xylose disodium salt
Formula: C₁₄H₂₂N₂Na₂O₁₆P₂
MW: 582.25 g/mol
Storage Temperature: Freezer
MDL Number: MFCD15145136
CAS Number: 108320-89-4

Order Now

Specification Test Results

Purity Limit 97%
Appearance White solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR