Fmoc-​Val-​Ala-​PAB-PNP 95%

Supplier: AFG Bioscience

C56032-100MG C56032-250MG C56032-500MG
76620-222EA 233.65 CAD
76620-222 76620-224 76620-226
Fmoc-​Val-​Ala-​PAB-PNP 95%
Fmoc-​Val-​Ala-​PAB-PNP
Formula: C₃₇H₃₆N₄O₉
Storage Temperature: Refrigerator
MDL Number: MFCD28557283
CAS Number: 1394238-92-6

Order Now

Specification Test Results

Purity Limit 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR