Disodium UDP-glucose ≥97%

Supplier: AFG Bioscience

C55202-1G C55202-2G C55202-500MG
76620-180EA 233.65 CAD
76620-180 76620-182 76620-178
Disodium UDP-glucose ≥97%
Disodium UDP-glucose
Formula: C₁₅H₂₂N₂Na₂O₁₇P₂
Storage Temperature: Freezer
MDL Number: MFCD00077895
CAS Number: 28053-08-9

Order Now

Specification Test Results

Purity Limit > 97%
Appearance White Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR