2-Deoxy-2-fluoro-L-fucose 95%

Supplier: AFG Bioscience

C52516-50MG C52516-100MG C52516-25MG
76619-812EA 949.44 CAD
76619-812 76619-814 76619-810
2-Deoxy-2-fluoro-L-fucose 95%
2-Deoxy-2-fluoro-L-fucose
Formula: C₆H₁₁FO₄
Storage Temperature: Freezer
MDL Number: MFCD08457748
CAS Number: 70763-62-1

Order Now

Specification Test Results

Purity Limit 95%
Appearance White solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR