α-D-Mannopyranosyl azide 95%

Supplier: AFG BIOSCIENCE LLC

Synonyms: 1-Azido-1-deoxy-α-D-mannose

C51542-100MG C51542-250MG C51542-500MG
76619-548EA 237.36 CAD
76619-548 76619-550 76619-552
α-D-Mannopyranosyl azide 95%
α-D-Mannopyranosyl azide
Formula: C₆H₁₁N₃O₅
MW: 205.169 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05865236
CAS Number: 51970-29-7

Order Now

Specification Test Results

Purity Limit 95%
Appearance White Crystalline Solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR