Benzyl 2-acetamido-2-deoxy-3-O-(β-D-galactopyranosyl)-α-D-galactopyranoside ≥95%

Supplier: AFG Bioscience

C54721-100MG C54721-25MG C54721-50MG
76620-138EA 1424.16 CAD
76620-138 76620-134 76620-136
Benzyl 2-acetamido-2-deoxy-3-O-(β-D-galactopyranosyl)-α-D-galactopyranoside ≥95%
Benzyl 2-acetamido-2-deoxy-3-O-(β-D-galactopyranosyl)-α-D-galactopyranoside
Formula: C₂₁H₃₁NO₁₁
Storage Temperature: Freezer
MDL Number: MFCD00057916
CAS Number: 3554-96-9

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR