4-Aminophenyl α-D-mannopyranoside ≥95%

Supplier: AFG Bioscience

C53753-250MG C53753-500MG
76619-966EA 474.73 CAD
76619-966 76619-968
4-Aminophenyl α-D-mannopyranoside ≥95%
4-Aminophenyl α-D-mannopyranoside
Formula: C₁₂H₁₇NO₆
Storage Temperature: Freezer
MDL Number: MFCD00067366
CAS Number: 34213-86-0

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR