2-Azidoethyl 2-acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranoside 95%

Supplier: AFG BIOSCIENCE LLC

Synonyms: 2-Azidoethyl 3,4,6-tri-O-acetyl-β-D-GlcNAc, 2-Azidoethyl 2-acetamido-2-deoxy-β-D-glucopyranoside 3,4,6-triacetate, 2-Azidoethyl 3,4,6-tri-O-acetyl-2-(acetylamino)-2-deoxy-β-D-glucopyranoside

C51595-1G C51595-250MG C51595-500MG
76619-626EA 1168.24 CAD
76619-626 76619-622 76619-624
2-Azidoethyl 2-acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranoside 95%
2-Azidoethyl 2-acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranoside
Formula: C₁₆H₂₄N₄O₉
MW: 416.383 g/mol
Storage Temperature: Freezer
MDL Number: MFCD19981038
CAS Number: 142072-11-5

Order Now

Specification Test Results

Purity Limit 95%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR