3,4,6-Tri-O-benzyl-1,2-O-(1-methoxyethylidene)-β-D-mannopyranose ≥95%

Supplier: AFG Bioscience

C53501-10G C53501-1G C53501-5G
76619-922EA 1557.64 CAD
76619-922 76619-918 76619-920
3,4,6-Tri-O-benzyl-1,2-O-(1-methoxyethylidene)-β-D-mannopyranose ≥95%
3,4,6-Tri-O-benzyl-1,2-O-(1-methoxyethylidene)-β-D-mannopyranose
Formula: C₃₀H₃₄O₇
Storage Temperature: Refrigerator
MDL Number: MFCD00270088
CAS Number: 16697-49-7

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White To Off-White Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR