4-Methylphenyl 1-thio-β-D-glucopyranoside 98%

Supplier: AFG BIOSCIENCE LLC

Synonyms: Tolyl 1-thio-β-D-glucopyranoside, Thiocresyl β-D-glucopyranoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(p-tolylthio)tetrahydro-2H-pyran-3,4,5-triol

C54673-10G C54673-1G C54673-5G
76620-120EA 1024.85 CAD
76620-120 76620-116 76620-118
4-Methylphenyl 1-thio-β-D-glucopyranoside 98%
4-Methylphenyl 1-thio-β-D-glucopyranoside
Formula: C₁₃H₁₈O₅S
MW: 286.344 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11045365
CAS Number: 1152-39-2

Order Now

Specification Test Results

Purity Limit 98%
Appearance White powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR