1-Azido-1-deoxy-β-D-glucopyranoside 97%

Supplier: AFG Bioscience

C51532-1G C51532-2G C51532-500MG
76619-520EA 632.96 CAD
76619-520 76619-522 77888-727
1-Azido-1-deoxy-β-D-glucopyranoside 97%
1-Azido-1-deoxy-β-D-glucopyranoside
Formula: C₆H₁₁N₃O₅
Storage Temperature: Refrigerator
MDL Number: MFCD03701503
CAS Number: 20379-59-3

Order Now

Specification Test Results

Purity Limit 97%
Appearance White Crystalline Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR