Acetaminophen-O-β-D-glucuronide sodium salt ≥95%

Supplier: AFG Bioscience

C54156-100MG C54156-250MG C54156-50MG
76620-048EA 395.6 CAD
76620-048 76620-050 76620-046
Acetaminophen-O-β-D-glucuronide sodium salt ≥95%
Acetaminophen-O-β-D-glucuronide sodium salt
Formula: C₁₄H₁₆NO₈Na
Storage Temperature: Freezer
MDL Number: MFCD00153898
CAS Number: 120595-80-4

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White To Off-White Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR