6-Azido-6-deoxy-D-galactopyranose 97%

Supplier: AFG Bioscience

Synonyms: 6-Azido-6-deoxy-D-galactose, (2R,3S,4S,5R)-6-Azido-2,3,4,5-tetrahydroxyhexanal

C51523-100MG C51523-250MG C51523-500MG
76619-484EA 236.75 CAD
76619-484 76619-486 76619-488
6-Azido-6-deoxy-D-galactopyranose 97%
6-Azido-6-deoxy-D-galactopyranose
Formula: C₆H₁₁N₃O₅
Storage Temperature: Refrigerator
MDL Number: MFCD03265529
CAS Number: 66927-03-5

Order Now

Specification Test Results

Purity Limit 97%
Appearance White Crystalline Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR