4-Aminophenyl beta-D-Galactopyranoside ≥95%
Supplier: AFG BIOSCIENCE LLC
Synonyms:
(2S,3R,4S,5R,6R)-2-(4-Aminophenoxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
C53747-1G
C53747-2G
C53747-5G
76619-956EA
233.65
CAD
76619-956
76619-958
76619-960
4-Aminophenyl beta-D-Galactopyranoside ≥95%
4-Aminophenyl beta-D-Galactopyranoside
Formula:
C₁₂H₁₇NO₆ MW: 271.27 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00067362 CAS Number: 5094-33-7 |
Specification Test Results
Purity Limit | > 95% |
Appearance | White Powder |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...