Azidoacetic acid 95-98%

Supplier: AFG Bioscience

C56672-10G C56672-1G C56672-5G
76620-286EA 631.73 CAD
76620-286 76620-282 76620-284
Azidoacetic acid 95-98%
Azidoacetic acid
Formula: C₂H₃N₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD01320907
CAS Number: 18523-48-3

Order Now

Specification Test Results

Purity Limit 95-98%
Appearance colorless liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR