2-NBDG ≥97%

Supplier: AFG Bioscience

C52303-10MG C52303-5MG
76619-794EA 411.43 CAD
76619-794 76619-792
2-NBDG ≥97%
2-NBDG
Formula: C₁₂H₁₄N₄O₈
Storage Temperature: Freezer
MDL Number: MFCD23135882
CAS Number: 186689-07-6

Order Now

Specification Test Results

Purity Limit > 97%
Appearance Crystalline Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR