3-Deazaadenosine 95%

Supplier: AFG Bioscience

Synonyms: c3Ado, 1-​β-​D-​ribofuranosyl-​1H-​Imidazo[4,​5-​c]pyridin-​4-​amine, deaza-Ado, 3-DZA, 4-Amino-1-(β-D-ribofuranosyl)-1H-imidazo(4,5)-pyridine

C55008-10MG C55008-5MG
76620-168EA 474.73 CAD
76620-168 76620-166
3-Deazaadenosine 95%
3-Deazaadenosine
Formula: C₁₁H₁₄N₄O₄
Storage Temperature: Freezer
MDL Number: MFCD00153951
CAS Number: 6736-58-9

Order Now

Specification Test Results

Purity Limit 95%
Appearance White solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR