D-Xylulose 98%

Supplier: AFG BIOSCIENCE LLC

Synonyms: D-threo-pent-2-uloseD-threo-2-pentulose

C50710-100MG C50710-250MG
76619-164EA 425.16 CAD
76619-164 76619-166
D-Xylulose 98%
D-Xylulose
Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00065958
CAS Number: 551-84-8

Order Now

Specification Test Results

Purity Limit 98%
Appearance Syrup

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR