Fmoc-Thr(GalNAc(Ac)₃-α-D)-OH ≥95%
Supplier: AFG BIOSCIENCE LLC
Synonyms:
N-(((9H-Fluoren-9-yl)methoxy)carbonyl)-O-((2S,3R,4R,5R,6R)-3-acetamido-4,5-diacetoxy-6-(acetoxymethyl)tetrahydro-2H-pyran-2-yl)-L-threonine, Fmoc-Thr[GalNAc(Ac)3-α-D]-OH, Fmoc-L-Thr(α-D-GalNAc(Ac)3)-OH
C51021-100MG
C51021-250MG
C51021-25MG
76619-322EA
934.59
CAD
76619-322
76619-324
76619-320
Fmoc-Thr(GalNAc(Ac)₃-α-D)-OH ≥95%
Fmoc-Thr(GalNAc(Ac)₃-α-D)-OH
Formula:
C₃₃H₃₈N₂O₁₃ MW: 670.67 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00171374 CAS Number: 116783-35-8 |
Specification Test Results
Purity Limit | > 95% |
Appearance | White Powder |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...