N-Acetyl-2,3-didehydro-2-deoxyneuraminic acid ≥95%
Supplier: AFG Bioscience
C53830-100MG
C53830-25MG
C53830-10MG
76619-974EA
1424.16
CAD
76619-974
76619-972
76619-970
N-Acetyl-2,3-didehydro-2-deoxyneuraminic acid ≥95%
N-Acetyl-2,3-didehydro-2-deoxyneuraminic acid
Formula:
C₁₁H₁₇NO₈ Storage Temperature: Freezer |
MDL Number:
MFCD00057470 CAS Number: 24967-27-9 |
Specification Test Results
Purity Limit | > 95% |
Appearance | White Powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...