Biotin-PEG3-azide 95%
Supplier: AFG BIOSCIENCE LLC
Synonyms:
N-[2-[2-[2-(2-Azidoethoxy)ethoxy]ethoxy]ethyl]biotinamide, Azido-PEG3-biotin conjugate
C56473-100MG
C56473-250MG
C56473-50MG
76620-270EA
395.6
CAD
76620-270
76620-272
76620-268
Biotin-PEG3-azide 95%
Biotin-PEG3-azide
Formula:
C₁₈H₃₂N₆O₅S MW: 444.55 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD20134145 CAS Number: 875770-34-6 |
Specification Test Results
Purity Limit | 95% |
Appearance | White powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...