2-Azidoethyl-2-acetamido-2-deoxy-β-D-glucopyranoside 95%

Supplier: AFG Bioscience

C51507-100MG C51507-250MG
76619-416EA 395.6 CAD
76619-416 76619-418
2-Azidoethyl-2-acetamido-2-deoxy-β-D-glucopyranoside 95%
2-Azidoethyl-2-acetamido-2-deoxy-β-D-glucopyranoside
Formula: C₁₀H₁₈N₄O₆
Storage Temperature: Freezer
CAS Number: 142072-12-6

Order Now

Specification Test Results

Purity Limit 95%
Appearance White powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR