11-Azido-3,6,9-trioxaundecan-1-amine 95%

Supplier: AFG Bioscience

Synonyms: 1-Amino-11-azido-3,6,9-trioxaundecane, Amino-PEG3-Azide, 11-Amino-3,6,9-trioxaundecanyl Azide, Azido-PEG3-Amine

C56450-1G C56450-5G
76620-258EA 155.76 CAD
76620-258 76620-260
11-Azido-3,6,9-trioxaundecan-1-amine 95%
11-Azido-3,6,9-trioxaundecan-1-amine
Formula: C₈H₁₈N₄O₃
MW: 218.26 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00269874
CAS Number: 134179-38-7

Order Now

Specification Test Results

Purity Limit 95%
Appearance colorless oil


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR