Mannose triflate ≥95%

Supplier: AFG Bioscience

C50648-500MG C50648-1G C50648-2G
76619-110EA 237.36 CAD
76619-110 76619-112 76619-114
Mannose triflate ≥95%
Formula: C₁₅H₁₉F₃O₁₂S
Storage Temperature: Freezer
MDL Number: MFCD00012353
CAS Number: 92051-23-5

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR