1,2,3,4-Tetra-O-acetyl-ɑ-L-fucopyranose ≥95%

Supplier: AFG Bioscience

C54827-2G C54827-5G C54827-10G
76620-152EA 316.47 CAD
76620-152 76620-154 76620-156
1,2,3,4-Tetra-O-acetyl-ɑ-L-fucopyranose ≥95%
1,2,3,4-Tetra-O-acetyl-alpha-L-fucopyranose
Formula: C₁₄H₂₀O₉
Storage Temperature: Refrigerator
MDL Number: MFCD00069791
CAS Number: 64913-16-2

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR