Isopropyl-β-D-thioglucopyranoside ≥97%

Supplier: AFG Bioscience

C54356-1G C54356-2G C54356-500MG
76620-078EA 514.27 CAD
76620-078 76620-080 76620-076
Isopropyl-β-D-thioglucopyranoside ≥97%
Isopropyl β-D-thioglucopyranoside
Formula: C₉H₁₈O₅S
Storage Temperature: Refrigerator
CAS Number: 19165-11-8

Order Now

Specification Test Results

Purity Limit > 97%
Appearance White Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR