Octyl-α-D-glucopyranoside ≥97%

Supplier: AFG BIOSCIENCE LLC

Synonyms: (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(octyloxy)tetrahydro-2H-pyran-3,4,5-triol, n-Octyl α-glucoside, Octyl α-D-glucopyranoside

C54386-1G C54386-5G
76620-082EA 395.6 CAD
76620-082 76620-084
Octyl-α-D-glucopyranoside ≥97%
Octyl α-D-glucopyranoside
Formula: C₁₄H₂₈O₆
MW: 292.37 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00070008
CAS Number: 29781-80-4

Order Now

Specification Test Results

Purity Limit > 97%
Appearance White Powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR