2-Deoxy-2-fluoro-D-glucose ≥98%

Supplier: AFG BIOSCIENCE LLC

Synonyms: 2-Fluoro-2-deoxy-D-glucose, 2-Deoxy-2-fluoro-D-glucopyranose, FDG

C52568-100MG C52568-250MG C52568-500MG
76619-822EA 316.47 CAD
76619-822 76619-824 76619-826
2-Deoxy-2-fluoro-D-glucose ≥98%
2-Deoxy-2-fluoro-D-glucose
Formula: C₆H₁₁FO₅
MW: 182.15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00077527
CAS Number: 29702-43-0

Order Now

Specification Test Results

Purity Limit > 98%
Appearance White Powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR