Nonyl-β-D-glucopyranoside 97%
Supplier: AFG Bioscience
Synonyms:
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-nonoxyoxane-3,4,5-triol, N-Nonyl Beta-D-glucopyranoside, β-Nonylglucoside, Nonyl β-D-glucopyranoside, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(nonyloxy)tetrahydro-2H-pyran-3,4,5-triol, NG
C54389-10G
C54389-1G
C54389-5G
76620-090EA
1345.05
CAD
76620-090
76620-086
76620-088
Nonyl-β-D-glucopyranoside 97%
Nonyl β-D-glucopyranoside
Formula:
C₁₅H₃₀O₆ MW: 306.4 Storage Temperature: Refrigerator |
MDL Number:
MFCD00063300 CAS Number: 69984-73-2 |
Specification Test Results
Purity Limit | 97% |
Appearance | White powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...