Dodecyl-β-D-glucopyranoside ≥97%

Supplier: AFG Bioscience

C54415-10G C54415-5G
76620-098EA 778.81 CAD
76620-098 76620-096
Dodecyl-β-D-glucopyranoside ≥97%
Dodecyl β-D-glucopyranoside
Formula: C₁₈H₃₆O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00063298
CAS Number: 59122-55-3

Order Now

Specification Test Results

Purity Limit > 97%
Appearance White Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR