Decyl-β-D-glucopyranoside ≥97%

Supplier: AFG BIOSCIENCE LLC

Synonyms: (2R,3R,4S,5S,6R)-2-(Decyloxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, N-Decyl beta-D-glucopyranoside, Decyl β-D-glucopyranoside, Decyl glucopyranoside, DG, Decyl beta-D-glucopyranoside

C54399-5G C54399-10G
76620-092EA 553.84 CAD
76620-092 76620-094
Decyl-β-D-glucopyranoside ≥97%
Decyl β-D-glucopyranoside
Formula: C₁₆H₃₂O₆
MW: 320.43 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00063297
CAS Number: 58846-77-8

Order Now

Specification Test Results

Purity Limit > 97%
Appearance White Powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR