Sterile SatPax® MicroSeal®-VP Pre-wetted Sealed Edge Wipes, 100% Polyester Knit
Supplier: Berkshire
SSPVPR00124 SSPVP00124
76635-706CS 1355.25 CAD
76635-706 77581-280
Sterile SatPax® MicroSeal®-VP Pre-wetted Sealed Edge Wipes, 100% Polyester Knit
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Sterile SatPax® MicroSeal®-VP is an ultrasonically sealed edge cleanroom laundered wipe recommended for ISO Class 3 and above environments. This wipe is composed of 100% continuous filament polyester knit fabric.
  • Pre-wetted with 70% USP Grade IPA
  • Ultrasonically sealed edge for reduced fiber contamination
  • Laundered and packaged in Berkshire's ISO Class 4 cleanroom with lot number on each pack
  • Lightweight, high absorbency material design reduces actual cost in use compared to heavier weight products
  • Critically low particles, fibers, ions, and extractables
More Product Information