Entris® II Essential Precision Balances, NTEP-Certified, Sartorius
Supplier: Sartorius
BCE323-1NUS BCE623-1NUS BCE653-1NUS BCE622-1NUS BCE1202-1NUS BCE3202-1NUS BCE4202-1NUS BCE5201-1NUS BCE6202-1NUS BCE8201-1NUS BCE323-1NUS BCE623-1NUS BCE653-1NUS BCE622-1NUS BCE1202-1NUS BCE3202-1NUS BCE4202-1NUS BCE5201-1NUS BCE6202-1NUS BCE8201-1NUS
76539-102EA 2627.94 CAD
76539-102 76539-104 76539-106 76539-108 76539-110 76539-112 76539-114 76539-116 76539-118 76539-120 CA76539-102 CA76539-104 CA76539-106 CA76539-108 CA76539-110 CA76539-112 CA76539-114 CA76539-116 CA76539-118 CA76539-120
Entris® II Essential Precision Balances, NTEP-Certified, Sartorius
Balances and Scales
100% NTEP-compliant/legal for trade weighing-Entris® II Essential line is the only precision balance in its class offering LED touch technology and 12 built-in applications.
  • Easy clean for operational efficiency and durability
  • Effective draft shield
  • Highest quality standards
  • Multilanguage (English, German, France, Italian, Spanish, Portuguese, Russian, Polish)
High chemical resistance ensured using parts made from hard-wearing polybutylene terephthalate (PBT), stainless steel and glass - prevents cross contamination with wipe clean design and easy-to-More Product Information