Entris® II Essential Analytical Balances, NTEP-Certified, Sartorius
Supplier: Sartorius
BCE124-1NUS BCE224-1NUS BCE124-1NUS BCE224-1NUS
76539-098EA 4459.34 CAD
76539-098 76539-100 CA76539-098 CA76539-100
Entris® II Essential Analytical Balances, NTEP-Certified, Sartorius
Balances and Scales
100% NTEP-compliant/legal for trade weighing-Entris® II Essential line is the only analytical balance in its class offering LED touch technology and 12 built-in applications.
  • Easy clean for operational efficiency and durability
  • Effective draft shield
  • Highest quality standards
  • Multilanguage (English, German, France, Italian, Spanish, Portuguese, Russian, Polish)
High chemical resistance ensured using parts made from hard-wearing polybutylene terephthalate (PBT), stainless steel and glass - prevents cross contamination with wipe clean design and easy-to-remove parts. Effective draft shield minimizes weighing errors caused by electrostatically charged samples using specially coated glass parts.
Automatically detects sartorius accessories (e.g. printer, second display). Real 'PC-direct feature' for easy connection to a PC to transfer weighing data directly into spreadsheets or documents such as Microsoft® Excel or Word.
Weighing chamber height: 240 mm