Boc-Val(D8)-Cit-PAB-OH ≥95%
Supplier: AFG Bioscience
C56093-100MG
C56093-25MG
76620-256EA
1323.99
CAD
76620-256
76620-254
Boc-Val(D8)-Cit-PAB-OH ≥95%
Boc-Val(D8)-Cit-PAB-OH
Specification Test Results
Purity Limit | >95% |
Appearance | Off-white powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...