Boc-Val(D8)-Cit-OH ≥95%
Supplier: AFG Bioscience
C56091-100MG
C56091-250MG
76620-246EA
700.94
CAD
76620-246
76620-248
Boc-Val(D8)-Cit-OH ≥95%
Boc-Val(D8)-Cit-OH
Specification Test Results
Purity Limit | >95% |
Appearance | Off-white powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...