Fmoc-Ala-Ala-PAB-PNP 95%

Supplier: AFG Bioscience

C56090-100MG C56090-250MG C56090-500MG
76620-240EA 395.6 CAD
76620-240 76620-242 76620-244
Fmoc-Ala-Ala-PAB-PNP 95%
Fmoc-Ala-Ala-PAB-PNP

Order Now

Specification Test Results

Purity Limit 95%
Appearance Off-white powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR