2-Acetamido-2,6-dideoxy-L-talose ≥95%
Supplier: AFG Bioscience
C50742-10MG
C50742-5MG
76619-248EA
1557.64
CAD
76619-248
76619-246
2-Acetamido-2,6-dideoxy-L-talose ≥95%
2-Acetamido-2,6-dideoxy-L-talose
Specification Test Results
Purity Limit | > 95% |
Appearance | White powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...