Racked Pipette Tips for Universal Fit Pipettes, Polycarbin
Supplier: POLYCARBIN, INC.
PC01-04-04 PC01-06-03 PC01-06-04 PC01-07-03 PC01-07-04
76635-516CS 531.54 CAD
76635-516 76635-500 76635-486 76635-478 76635-514 76635-476 76635-496 76635-504
Racked Pipette Tips for Universal Fit Pipettes, Polycarbin
Pipette Tips
Polycarbin's pipette tip rack is made of 100% circular economy, post-consumer recycled content, combining high performance with a significantly reduced carbon footprint. Pipette tips do not contain recycled content, and are universal fit.
  • Rack made from 100% circular economy, recycled content
  • Manufactured in an ISO 9001 compliant facility
  • Carries a smaller carbon footprint
  • DNase, RNase and Pyrogen free
  • Sterile
Polycarbin's line of low-carbon lab products are designed for the sustainability-minded scientist. When designing our line of pipette tips, weMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Sustainable Packaging,Waste Reducing,Product Transparency and Disclosure Sustainable Materials,Safer Human and Environmental Health,Low Manufacturing Impact,Energy Efficient,Waste Reducing,Product Transparency and Disclosure,Water Efficient
Environmental certification This environmental impact of this product was evaulated by a third party, panel reviewed LCA to determine recycled content, carbon dioxide emission eq. reduction, water conservation, and crude oil conservation.