PUREZERO* HG5 Blue SGX* Non Sterile Nitrile Gloves, Halyard
Supplier: O&M HALYARD CANADA, INC CA
CLN92512LG CLN92512MD CLN92512SM CLN92512XL CLN92512XS
76635-368CS 455.97 CAD
76635-368 76635-366 76635-364 76635-370 76635-362
PUREZERO* HG5 Blue SGX* Non Sterile Nitrile Gloves, Halyard
Gloves
Halyard PUREZERO* HG5 blue SGX* nitrile cleanroom gloves are designed for semiconductor, pharmaceutical, and medical device manufacturing applications. They are recommended for use in ISO Class 5 or higher and Grade B/C/D cleanrooms.
  • Ambidextrous design
  • Smooth Grip
  • AQL 1.0
  • Latex-free, powder-free and silicone-free
  • Low dermatitis potential
  • Static dissipative in use
  • Max particle count: 2000
PUREZERO* HG5 blue SGX* nitrile cleanroom gloves are clean processed (washed repeatedly in deionized water) to ensure consistent control of low particles andMore Product Information