PUREZERO* HG5 White Non Sterile Nitrile Gloves
Supplier: O&M HALYARD CANADA, INC CA
CLN30512LG CLN30512MD CLN30512SM CLN30512XL CLN30512XS CLN305122XL
76635-378CS 364.71 CAD
76635-378 76635-376 76635-374 76635-380 76635-372 77575-304
PUREZERO* HG5 White Non Sterile Nitrile Gloves
Gloves
Halyard PUREZERO* HG5 White Nitrile Cleanroom Gloves are designed for semiconductor, pharmaceutical and medical device manufacturing applications. They are recommended for use in ISO Class 5 or higher and grade B/C/D cleanrooms.
  • Ambidextrous design
  • Textured fingertips, with tacky grip
  • AQL 1.0
  • Latex-free, powder-free and silicone-free
  • Low dermatitis potential
  • Static dissipative in use
  • Max particle count: 1500
PUREZERO* HG5 White Nitrile Cleanroom Gloves are clean processed (washed repeatedly in deionized water) to ensure consistent control of lowMore Product Information
New Product
Order Now
SPECIFICATIONS
Ambidextrous/Hand-specific Ambidextrous
Environmentally Preferable
Latex/Latex-free Latex-free
Powdered/Powder-free Powder-free
Material Nitrile
Finish Tacky grip
Sterility Non sterile