Paraformaldehyde 8% (w/v) in aqueous solution methanol-free

Supplier: Thermo Scientific Chemicals
Danger

047347-9L 047347-9M
AA47347-9LEA 62.19 CAD
AA47347-9L AA47347-9M
Paraformaldehyde 8% (w/v) in aqueous solution methanol-free
Formaldehyde

Solution is micro-filtered and supplied in ampules. RNase and DNase free.

Formula: CH₂O
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0

Order Now

Specification Test Results

Concentration (w/v) 7.5 to 8%
Appearance Clear colourless liquid
Ash <=0.01% (Paraformaldehyde content)
Trace metal
Iron (Fe) <=2.0% (Paraformaldehyde content)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR