Paraformaldehyde 10% (w/v) in aqueous solution methanol-free

Supplier: Thermo Scientific Chemicals
Danger

047317-9L 047317-9M
AA47317-9LEA 66.68 CAD
AA47317-9L AA47317-9M
Paraformaldehyde 10% (w/v) in aqueous solution methanol-free
Formaldehyde

Solution is micro-filtered and supplied in ampules. RNase and DNase free.

Formula: CH₂O
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
UN: 3334
ADR: 9,III

Order Now

Specification Test Results

Concentration (w/v) 9.5 to 10%
Appearance Clear colourless liquid
Ash <=0.01% (Paraformaldehyde content)
Trace metal
Iron (Fe) <=2.0% (Paraformaldehyde content)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR