Biohazard Disposable Bags, PP
Supplier: United Scientific Supplies
UNU40106 UNU40103 UNU40104 UNU40105
470347-982PK 123.85 CAD
470347-982 470347-970 470347-912 470347-948 470347-942
Biohazard Disposable Bags, PP
Bags Waste Disposal Bags
Biohazard disposal bags are ideal for processing biohazardous materials that require autoclaving prior to disposal.
  • Polypropylene bags include black biohazard symbol and precautionary procedures printed in four languages (English, Spanish, French, German) for added safety
  • High temperature steam autoclavable to 121 °C (250 °F)
  • Pass ASTM 1922 Tear Resistance and ASTM 1709 Dart Impact Tests
  • 55 microns (2.2 mil) thickness
These bright red, easy to open, polypropylene (PP) Biohazard Bags are for disposal of biohazard waste and plastic laboratory items such as used tips and tubes. Bags have a clearly printed biohazard symbol and pass ASTM 1922 tear resistance and ASTM 1709 dart impact tests.
Steam autoclavable for sterilization of waste before disposal.