REUZ™ Stainless Steel Beakers with Handle
Supplier: United Scientific Supplies
UN3003-1000 UN3003-250 UN3003-500
470347-966EA 68.35 CAD
470347-966 470347-918 470347-962
REUZ™ Stainless Steel Beakers with Handle
Beakers
Stainless steel beakers with convenient handles.
  • 304 stainless steel
  • Pouring spout and large handle
  • Easy to clean
  • Impact-resistant
  • Available in 250, 500 and 1000 ml sizes
Manufactured from 304 stainless steel, REUZ™ ('re-use') beakers with handles feature graduations stamped into the inside wall for easy reading of volumes in milliliters or ounces. Include a pouring spout and large, comfortable stainless steel handle. The precision corrosion-resistant stainless construction includes rolled top rims, smooth edges and radiused corners for easy, thoroughMore Product Information