REUZ™ Stainless Steel Pharma Scoops
Supplier: United Scientific Supplies
UN5003-100 UN5003-1000 UN5003-1250 UN5003-1700 UN5003-2500 UN5003-750
470347-950EA 131.25 CAD
470347-950 470347-946 470347-926 470347-936 470347-920 470347-940
REUZ™ Stainless Steel Pharma Scoops
Scoops
Reusable stainless steel scoops for powders and granulated materials.
  • 304 stainless steel
  • Smooth, polished surface
  • Easy to clean
  • Large handle
  • Available in sizes from 10 to 2500 ml
Manufactured with 304 stainless steel, REUZ™ ('re-use') Pharma Scoops have polished surfaces and rounded edges. The precision corrosion-resistant stainless construction includes large handles and smooth, large radius interior surfaces for easy cleaning and protection against bacteria deposits. Includes large rims near the handle to protect against spills when the scoop is tilted up. Intended for handling powders and granules in laboratory and production settings.
REUZ™ products promote sustainability in all laboratory environments.
Ordering information: Sold individually.