REUZ™ Stainless Steel Scoops
Supplier: United Scientific Supplies
UN5002-100 UN5002-200 UN5002-250 UN5002-350 UN5002-500
470347-968EA 49.4 CAD
470347-968 470347-916 470347-978 470347-960 470347-922
REUZ™ Stainless Steel Scoops
Scoops
These reusable stainless steel scoops allow you to easily transfer powders and granular products.
  • 304 stainless steel
  • Smooth, polished surfaces
  • Easy to clean
  • Large handle
  • Available in sizes from 100 to 500 ml
Manufactured from 304 stainless steel, REUZ™ ('re-use') scoops are suitable for use with powders and granules in a variety of environments. Rounded bowls of the scoops are completely seamless for easy cleaning. The corrosion-resistant stainless construction includes large handles and smooth, large radius interior surfaces for easy cleaning and protection against bacteria deposits.
REUZ™ products promote sustainability in all laboratory environments.
Ordering information: Sold individually.